Product Details

SNP ID
rs28409868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103123908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGCCGGGCCCAGCGAGCTCC[C/G]GGGAGGTGTGTGGACATCAGCCCTG
Phenotype
MIM: 603300
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00677 PubMed Links

Gene Details

Gene
LINC00677
Gene Name
long intergenic non-protein coding RNA 677
There are no transcripts associated with this gene.

Gene
TNFAIP2
Gene Name
TNF alpha induced protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006291.2 581 Intron NP_006282.2
XM_006720243.3 581 UTR 5 XP_006720306.1
XM_011537112.2 581 UTR 5 XP_011535414.1
XM_011537113.2 581 Intron XP_011535415.1
XM_011537114.2 581 UTR 5 XP_011535416.1
XM_011537115.2 581 Missense Mutation CCG,CGG P240R XP_011535417.1
XM_017021616.1 581 Intron XP_016877105.1

View Full Product Details