Product Details

SNP ID
rs34865723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104769973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAGCTGGTGACAGACAGCCCAG[G/T]GCGGCTGGCTGACAGAGTGAGGGGA
Phenotype
MIM: 164730
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKT1 PubMed Links

Gene Details

Gene
AKT1
Gene Name
AKT serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014431.1 2151 UTR 3 NP_001014431.1
NM_001014432.1 2151 UTR 3 NP_001014432.1
NM_005163.2 2151 UTR 3 NP_005154.2
XM_005267401.1 2151 UTR 3 XP_005267458.1
XM_017021075.1 2151 Intron XP_016876564.1
XM_017021076.1 2151 Intron XP_016876565.1
XM_017021077.1 2151 Intron XP_016876566.1
XM_017021078.1 2151 Intron XP_016876567.1

View Full Product Details