Product Details
- SNP ID
-
rs28678492
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:105405883 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGGAAAATAAGAGCAAGGCCTG[G/T]CTTGTCCGCACCAGCTCCAACAACT
- Phenotype
-
MIM: 610423
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PACS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79235548] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PACS2
- Gene Name
- phosphofurin acidic cluster sorting protein 2
There are no transcripts associated with this gene.
- Gene
- TEX22
- Gene Name
- testis expressed 22
View Full Product Details