Product Details
- SNP ID
-
rs28521273
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124008006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGAGAGAAAGGCCCCTGGAGGGA[A/G]AAACCATGTCTGGATGGCATTGAGT
- Phenotype
-
MIM: 608277
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHST15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79057288] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHST15
- Gene Name
- carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
View Full Product Details