Product Details

SNP ID
rs11550197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86959641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATTTCCTCCCCAGGAGCCAAG[A/C]CCAAGGAGAATGTTGTACAGAGCGT
Phenotype
MIM: 608925 MIM: 602998
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADIRF PubMed Links

Gene Details

Gene
ADIRF
Gene Name
adipogenesis regulatory factor
There are no transcripts associated with this gene.

Gene
MMRN2
Gene Name
multimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024756.2 297 Intron NP_079032.2
XM_005270153.1 297 Intron XP_005270210.1
XM_006717970.3 297 Intron XP_006718033.1
Gene
SNCG
Gene Name
synuclein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003087.2 297 Missense Mutation ACC,CCC T44P NP_003078.2
XM_011540105.2 297 Missense Mutation ACC,CCC T44P XP_011538407.1

View Full Product Details