Product Details
- SNP ID
-
rs28523711
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:48735515 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAGGAGGAAGGAAGGAGGGAAG[A/G]AAGGAAGGAAGTAAGGAAGGAAGAA
- Phenotype
-
MIM: 616476
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGBL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147008867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGBL4
- Gene Name
- ATP/GTP binding protein like 4
- Gene
- BEND5
- Gene Name
- BEN domain containing 5
View Full Product Details