Product Details

SNP ID
rs28602706
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:74734289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCTTTGTCCAGAATATTCCAC[C/T]ATATATTCTTGCTGCCCTTTAGCTG
Phenotype
MIM: 123691 MIM: 611245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 Intron NP_001123514.1
NM_001130043.1 Intron NP_001123515.1
NM_001134759.1 Intron NP_001128231.1
NM_001889.3 Intron NP_001880.2
XM_005270491.4 Intron XP_005270548.1
XM_011540747.1 Intron XP_011539049.1
XM_017000367.1 Intron XP_016855856.1
XM_017000368.1 Intron XP_016855857.1
Gene
TYW3
Gene Name
tRNA-yW synthesizing protein 3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162916.1 Intron NP_001156388.1
NM_138467.2 Intron NP_612476.1
XM_006710347.1 Intron XP_006710410.1

View Full Product Details