Product Details

SNP ID
rs35083316
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:77979512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGGCGAAGCAACAGTGTCTCGA[C/G]AGCGCTGCCGCCCGGAGGGCGCCAG
Phenotype
MIM: 611327 MIM: 603444
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB4 PubMed Links

Gene Details

Gene
DNAJB4
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317099.1 355 Intron NP_001304028.1
NM_001317100.1 355 UTR 5 NP_001304029.1
NM_001317101.1 355 UTR 5 NP_001304030.1
NM_001317102.1 355 Intron NP_001304031.1
NM_001317103.1 355 Intron NP_001304032.1
NM_007034.4 355 Intron NP_008965.2
Gene
FUBP1
Gene Name
far upstream element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303433.1 355 Intron NP_001290362.1
NM_003902.4 355 Intron NP_003893.2
XM_011542391.2 355 Intron XP_011540693.1
XM_011542392.2 355 Intron XP_011540694.1
XM_011542393.2 355 Intron XP_011540695.1
XM_017002739.1 355 Intron XP_016858228.1
XM_017002740.1 355 Intron XP_016858229.1
XM_017002741.1 355 Intron XP_016858230.1
XM_017002742.1 355 Intron XP_016858231.1
XM_017002743.1 355 Intron XP_016858232.1
XM_017002744.1 355 Intron XP_016858233.1

View Full Product Details