Product Details

SNP ID
rs35231465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102713404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGCAACTCTGGTAACTCTCT[A/G]AGCAATAAGATCAAATGCGTAATAT
Phenotype
MIM: 120355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
There are no transcripts associated with this gene.

Gene
MMP8
Gene Name
matrix metallopeptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304441.1 1527 Nonsense Mutation CAG,TAG Q427* NP_001291370.1
NM_001304442.1 1527 Nonsense Mutation CAG,TAG Q427* NP_001291371.1
NM_002424.2 1527 Nonsense Mutation CAG,TAG Q450* NP_002415.1
XM_011542834.2 1527 Nonsense Mutation CAG,TAG Q459* XP_011541136.1
XM_011542835.2 1527 Nonsense Mutation CAG,TAG Q427* XP_011541137.1
XM_011542836.2 1527 UTR 3 XP_011541138.1
XM_017017771.1 1527 Nonsense Mutation CAG,TAG Q427* XP_016873260.1

View Full Product Details