Product Details

SNP ID
rs35024560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68315020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATTTAGTAGAACTCACACACAC[A/G]GGTGGGGGATGTAGGCAGGTGAGGC
Phenotype
MIM: 603506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP5 PubMed Links

Gene Details

Gene
LRP5
Gene Name
LDL receptor related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291902.1 Intron NP_001278831.1
NM_002335.3 Intron NP_002326.2
XM_005273994.2 Intron XP_005274051.1
XM_011545029.1 Intron XP_011543331.1
XM_011545030.1 Intron XP_011543332.1
XM_011545031.1 Intron XP_011543333.1
XM_017017735.1 Intron XP_016873224.1
XM_017017736.1 Intron XP_016873225.1

View Full Product Details