Product Details

SNP ID
rs28362943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57598160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCGCTAAGAGGGACTGGGGCC[G/T]GAGACGGAATGGGGGCGGGCCCCGG
Phenotype
MIM: 606860
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPING1 PubMed Links

Gene Details

Gene
SERPING1
Gene Name
serpin family G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000062.2 Intron NP_000053.2
NM_001032295.1 Intron NP_001027466.1

View Full Product Details