Product Details
- SNP ID
-
rs28710163
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:134140440 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGGTGGATTGAGGGGGGCCTCA[C/T]GTTTCTGGCTCGGCTGGTGCTGTTG
- Phenotype
-
MIM: 609012
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR5
PubMed Links
Gene Details
- Gene
- WDR5
- Gene Name
- WD repeat domain 5
View Full Product Details