Product Details

SNP ID
rs28538762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100553719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAACCACCGATGGCGTTTCACA[A/T]CATTCGCCCCGTTCTTCATATTTCC
Phenotype
MIM: 615276
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CERS3 PubMed Links

Gene Details

Gene
CERS3
Gene Name
ceramide synthase 3
There are no transcripts associated with this gene.

Gene
LOC102723335
Gene Name
uncharacterized LOC102723335
There are no transcripts associated with this gene.

Gene
PRKXP1
Gene Name
protein kinase, X-linked, pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details