Product Details

SNP ID
rs28687624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101281899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACTGAAGTAGGTACAAAAAGA[A/T]CAAAAGCAGCACCTGCCTTTGAGGA
Phenotype
MIM: 603521 MIM: 607918
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRPA1 PubMed Links

Gene Details

Gene
SNRPA1
Gene Name
small nuclear ribonucleoprotein polypeptide A'
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003090.3 Intron NP_003081.2
Gene
VIMP
Gene Name
VCP interacting membrane selenoprotein
There are no transcripts associated with this gene.

View Full Product Details