Product Details

SNP ID
rs34226328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98649596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAAAGGAATGAAGTCTGGCTC[C/T]GGAGGAGGGTCCCCGACCTCGCTGT
Phenotype
MIM: 147370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGF1R PubMed Links

Gene Details

Gene
IGF1R
Gene Name
insulin like growth factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000875.4 1058 Silent Mutation TCC,TCT S5S NP_000866.1
NM_001291858.1 1058 Silent Mutation TCC,TCT S5S NP_001278787.1
XM_011521516.2 1058 Intron XP_011519818.1
XM_011521517.2 1058 Intron XP_011519819.1
XM_017022136.1 1058 Intron XP_016877625.1
XM_017022137.1 1058 Intron XP_016877626.1
XM_017022138.1 1058 Intron XP_016877627.1
XM_017022139.1 1058 Intron XP_016877628.1
Gene
IRAIN
Gene Name
IGF1R antisense imprinted non-protein coding RNA
There are no transcripts associated with this gene.

View Full Product Details