Product Details

SNP ID
rs28533314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:289082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGGCCGGAAGCCCTTTTTCTT[C/T]TTTTTTTTTTTTTGGGCGGGGGTAG
Phenotype
MIM: 602844 MIM: 603816 MIM: 608012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIG PubMed Links

Gene Details

Gene
ARHGDIG
Gene Name
Rho GDP dissociation inhibitor gamma
There are no transcripts associated with this gene.

Gene
AXIN1
Gene Name
axin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003502.3 Intron NP_003493.1
NM_181050.2 Intron NP_851393.1
XM_011522682.2 Intron XP_011520984.1
XM_011522683.2 Intron XP_011520985.1
XM_011522684.2 Intron XP_011520986.1
XM_011522686.1 Intron XP_011520988.1
XM_017023743.1 Intron XP_016879232.1
XM_017023744.1 Intron XP_016879233.1
XM_017023745.1 Intron XP_016879234.1
XM_017023746.1 Intron XP_016879235.1
XM_017023747.1 Intron XP_016879236.1
XM_017023748.1 Intron XP_016879237.1
Gene
PDIA2
Gene Name
protein disulfide isomerase family A member 2
There are no transcripts associated with this gene.

View Full Product Details