Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301108.1 | Intron | NP_001288037.1 | |||
NM_001301109.1 | Intron | NP_001288038.1 | |||
NM_001324182.1 | Intron | NP_001311111.1 | |||
NM_001324183.1 | Intron | NP_001311112.1 | |||
NM_001324184.1 | Intron | NP_001311113.1 | |||
NM_001324185.1 | Intron | NP_001311114.1 | |||
NM_001324186.1 | Intron | NP_001311115.1 | |||
NM_001324187.1 | Intron | NP_001311116.1 | |||
NM_003027.4 | Intron | NP_003018.3 | |||
XM_011521889.1 | Intron | XP_011520191.1 | |||
XM_011521891.2 | Intron | XP_011520193.1 | |||
XM_011521892.2 | Intron | XP_011520194.1 | |||
XM_017022486.1 | Intron | XP_016877975.1 |