Product Details
- SNP ID
-
hCV64444472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37780494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATGACAAATTGAAATTTTATA[C/T]CATTTTCTAAATAAAAAATATAAGA
- Phenotype
-
MIM: 300481
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYBB
PubMed Links
Gene Details
- Gene
- CYBB
- Gene Name
- cytochrome b-245 beta chain
There are no transcripts associated with this gene.
View Full Product Details