Product Details
- SNP ID
-
rs62001917
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:156041545 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGCTGCATGAGTTGCTGCTGGGC[A/G]CTGGAAGCCCCTGTTGGAGAAGATG
- Phenotype
-
MIM: 605440
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBQLN4
PubMed Links
Gene Details
- Gene
- UBQLN4
- Gene Name
- ubiquilin 4
View Full Product Details