Product Details
- SNP ID
-
rs61747610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9471414 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGCTTGACAGAGGCAGTCACC[A/G]TGGCAGGTGAGCAGTTGTGTTTGAA
- Phenotype
-
MIM: 603433
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF143
PubMed Links
Gene Details
- Gene
- ZNF143
- Gene Name
- zinc finger protein 143
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282656.1 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
NP_001269585.1 |
NM_001282657.1 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
NP_001269586.1 |
NM_003442.5 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
NP_003433.3 |
XM_005253121.1 |
225 |
Missense Mutation |
ATG,GTG |
M51V |
XP_005253178.1 |
XM_005253122.1 |
225 |
Missense Mutation |
ATG,GTG |
M51V |
XP_005253179.1 |
XM_005253127.1 |
225 |
UTR 5 |
|
|
XP_005253184.1 |
XM_011520349.2 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
XP_011518651.1 |
XM_017018252.1 |
225 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873741.1 |
XM_017018253.1 |
225 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873742.1 |
XM_017018254.1 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
XP_016873743.1 |
XM_017018255.1 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
XP_016873744.1 |
XM_017018256.1 |
225 |
Missense Mutation |
ATG,GTG |
M36V |
XP_016873745.1 |
View Full Product Details