Product Details

SNP ID
rs74033340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88653535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCAGGGCTCAGGGAGGGGGAGAC[A/G]GCAGGTGGCTGCTGTGGGGACAGGT
Phenotype
MIM: 608508 MIM: 603236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYBA PubMed Links

Gene Details

Gene
CYBA
Gene Name
cytochrome b-245 alpha chain
There are no transcripts associated with this gene.

Gene
MVD
Gene Name
mevalonate diphosphate decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002461.2 Intron NP_002452.1
XM_011523086.2 Intron XP_011521388.1
XM_011523087.2 Intron XP_011521389.1
XM_011523088.2 Intron XP_011521390.1
XM_011523089.2 Intron XP_011521391.1
Gene
SNAI3-AS1
Gene Name
SNAI3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details