Product Details

SNP ID
rs59497198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3197847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCATGATAGCCTTGGCCAAGG[A/C]AGACAGCTATACCTTGGCTGCAATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR1A2 PubMed Links

Gene Details

Gene
OR1A2
Gene Name
olfactory receptor family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012352.2 331 Missense Mutation GAA,GCA E110A NP_036484.1

View Full Product Details