Product Details

SNP ID
rs58239286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52841666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACATTCAAAAGATTTTTCTCTCA[G/T]GTGTACTTCCCATTTTTGTGTGAGT
Phenotype
MIM: 616841
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF468 PubMed Links

Gene Details

Gene
ZNF468
Gene Name
zinc finger protein 468
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008801.1 994 Intron NP_001008801.1
NM_001277120.1 994 Intron NP_001264049.1
XM_017027443.1 994 Missense Mutation ATG,CTG M229L XP_016882932.1
XM_017027444.1 994 Missense Mutation ATG,CTG M157L XP_016882933.1

View Full Product Details