Product Details

SNP ID
rs61744387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86206204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATTGTCCACTGGACGTTTTAGT[A/C]ATATTCAGACACCAGTTGTTTCCTC
Phenotype
MIM: 611841 MIM: 609139
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL35 PubMed Links

Gene Details

Gene
MRPL35
Gene Name
mitochondrial ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016622.3 200 Missense Mutation AAT,CAT N48H NP_057706.2
NM_145644.2 200 Missense Mutation AAT,CAT N48H NP_663619.1
Gene
REEP1
Gene Name
receptor accessory protein 1
There are no transcripts associated with this gene.

View Full Product Details