Product Details

SNP ID
rs41312635
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:42180041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCCAAAGTCACACAACCAGTA[A/G]GGGACAAAGATAGGATCTGAGGTCA
Phenotype
MIM: 616035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXJ3 PubMed Links

Gene Details

Gene
FOXJ3
Gene Name
forkhead box J3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198850.1 Intron NP_001185779.1
NM_001198851.1 Intron NP_001185780.1
NM_001198852.1 Intron NP_001185781.1
NM_014947.4 Intron NP_055762.3
XM_005270632.4 Intron XP_005270689.1
XM_006710458.3 Intron XP_006710521.1
XM_006710459.3 Intron XP_006710522.1
XM_011541026.2 Intron XP_011539328.1
XM_017000693.1 Intron XP_016856182.1
XM_017000694.1 Intron XP_016856183.1
XM_017000695.1 Intron XP_016856184.1

View Full Product Details