Product Details

SNP ID
rs144163296
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:25390838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAGAGCATAGTAGGTGTTGAACA[C/T]GGCATTCTTCCTTTGGATTGGACTT
Phenotype
MIM: 111700
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RHCE PubMed Links
Additional Information
For this assay, SNP(s) [rs1053361] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RHCE
Gene Name
Rh blood group CcEe antigens
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020485.4 798 Missense Mutation ATG,GTG M238V NP_065231.3
NM_138616.3 798 Intron NP_619522.3
NM_138617.3 798 Intron NP_619523.3
NM_138618.3 798 Missense Mutation ATG,GTG M238V NP_619524.3
XM_005245957.3 798 Missense Mutation ATG,GTG M238V XP_005246014.1
XM_006710810.3 798 Intron XP_006710873.1
XM_011541888.2 798 Missense Mutation ATG,GTG M198V XP_011540190.1
XM_011541889.2 798 Missense Mutation ATG,GTG M195V XP_011540191.1
XM_011541890.2 798 Missense Mutation ATG,GTG M238V XP_011540192.1
XM_011541891.2 798 Missense Mutation ATG,GTG M182V XP_011540193.1
XM_017002014.1 798 Missense Mutation ATG,GTG M238V XP_016857503.1

View Full Product Details