Product Details
- SNP ID
-
rs121909377
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47333238 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGCACTGTGTACCCCCAGAGCT[C/A]CGTGTTGCCGACATCCTGGGGTGGC
- Phenotype
-
MIM: 603584
MIM: 600958
- Polymorphism
- C/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MADD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1052373] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MADD
- Gene Name
- MAP kinase activating death domain
There are no transcripts associated with this gene.
- Gene
- MYBPC3
- Gene Name
- myosin binding protein C, cardiac
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000256.3 |
3341 |
Missense Mutation |
|
|
NP_000247.2 |
View Full Product Details