Product Details

SNP ID
rs41285796
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:946132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTGCGCTAGATCTGAAACCCAG[G/T]AAGTCACAACACACCCCCAGGTCCC
Phenotype
MIM: 610770 MIM: 616765
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOC2L PubMed Links

Gene Details

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015658.3 Intron NP_056473.2
Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
There are no transcripts associated with this gene.

View Full Product Details