Product Details

SNP ID
rs41263656
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:152970877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAGAAAGTTTAATCTCCATTCC[A/G]CACCATTGCTCAGTATGACTTATTT
Phenotype
MIM: 616363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 Intron NP_775103.1
XM_017000482.1 Intron XP_016855971.1

View Full Product Details