Product Details

SNP ID
rs45521041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222200634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCCACCGTGCCCTTCCTCCAA[C/G]CCAGGTGGGCTACCAAATAAATGTG
Phenotype
MIM: 606597
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAX3 PubMed Links

Gene Details

Gene
PAX3
Gene Name
paired box 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000438.5 2610 Intron NP_000429.2
NM_001127366.2 2610 UTR 3 NP_001120838.1
NM_013942.4 2610 Intron NP_039230.1
NM_181457.3 2610 Intron NP_852122.1
NM_181458.3 2610 UTR 3 NP_852123.1
NM_181459.3 2610 UTR 3 NP_852124.1
NM_181460.3 2610 UTR 3 NP_852125.1
NM_181461.3 2610 UTR 3 NP_852126.1

View Full Product Details