Product Details

SNP ID
rs45463801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128480473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCAGCTCACCCTCCCTGGGCC[A/G]TGGCCACTCTGGCTTTGGCTCAGCC
Phenotype
MIM: 137295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB8-AS1 PubMed Links

Gene Details

Gene
DNAJB8-AS1
Gene Name
DNAJB8 antisense RNA 1
There are no transcripts associated with this gene.

Gene
GATA2
Gene Name
GATA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145661.1 2323 UTR 3 NP_001139133.1
NM_001145662.1 2323 UTR 3 NP_001139134.1
NM_032638.4 2323 UTR 3 NP_116027.2
Gene
GATA2-AS1
Gene Name
GATA2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details