Product Details

SNP ID
rs41270557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:79916247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCAAAATAATTCATAAAGACC[A/G]TTTCTGTGATCGTCTAAAATTCAGA
Phenotype
MIM: 605512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELOVL4 PubMed Links

Gene Details

Gene
ELOVL4
Gene Name
ELOVL fatty acid elongase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022726.3 1625 UTR 3 NP_073563.1

View Full Product Details