Product Details

SNP ID
rs45474692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120274285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATATTATCCAGACTGTGCCGGG[A/G]AGAAATCAAAAACACCTGTTTGAAG
Phenotype
MIM: 605410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCND2 PubMed Links

Gene Details

Gene
KCND2
Gene Name
potassium voltage-gated channel subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012281.2 618 UTR 5 NP_036413.1
XM_011516165.2 618 UTR 5 XP_011514467.1

View Full Product Details