Product Details

SNP ID
rs45585037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112944550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAACAGTGGCTGCCAAGGGGGC[A/G]AGCATGCGGCCTGCTTCACAGAGCT
Phenotype
MIM: 603351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OAS3 PubMed Links

Gene Details

Gene
OAS3
Gene Name
2'-5'-oligoadenylate synthetase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006187.3 554 Missense Mutation AAG,GAG K179E NP_006178.2
XM_005253889.3 554 Missense Mutation AAG,GAG K179E XP_005253946.1
XM_017019363.1 554 Missense Mutation AAG,GAG K179E XP_016874852.1

View Full Product Details