Product Details

SNP ID
rs41300888
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107981311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGATCTTTCTCCCTCTGCCCC[A/G]CAAAGGTGTAATTTGCCTGCTACAA
Phenotype
MIM: 300313
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC105373310 PubMed Links

Gene Details

Gene
LOC105373310
Gene Name
NACHT, LRR and PYD domains-containing protein 10-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017030015.1 801 Intron XP_016885504.1
Gene
TEX13B
Gene Name
testis expressed 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031273.2 801 Silent Mutation TGC,TGT C236C NP_112563.1

View Full Product Details