Product Details

SNP ID
rs45557031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120626739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGATAGCAAACGTAGTGGGGCGT[A/G]CAAGGAAGAACCAGTTGTATTGGTC
Phenotype
MIM: 300611 MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1GALT1C1 PubMed Links

Gene Details

Gene
C1GALT1C1
Gene Name
C1GALT1 specific chaperone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011551.2 667 Missense Mutation GCA,GTA A143V NP_001011551.1
NM_152692.4 667 Missense Mutation GCA,GTA A143V NP_689905.1
Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
There are no transcripts associated with this gene.

View Full Product Details