Product Details

SNP ID
rs45489901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50255270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTGGACCTGGGGCTCATCTTC[C/T]GTGTCGTGCAGGGACTCGAAGTAGG
Phenotype
MIM: 608544 MIM: 602898 MIM: 602399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDAC10 PubMed Links

Gene Details

Gene
HDAC10
Gene Name
histone deacetylase 10
There are no transcripts associated with this gene.

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
There are no transcripts associated with this gene.

Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303252.1 1171 Silent Mutation NP_001290181.1
NM_002969.4 1171 Silent Mutation NP_002960.2

View Full Product Details