Product Details

SNP ID
rs45559634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8880815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGGTCTAACAAACAACTACAT[A/T]AACCTCGGGCAACAACCATCATGGA
Phenotype
MIM: 605256
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAD18 PubMed Links

Gene Details

Gene
RAD18
Gene Name
RAD18, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020165.3 2120 UTR 3 NP_064550.3
XM_017006873.1 2120 UTR 3 XP_016862362.1

View Full Product Details