Product Details

SNP ID
rs41307457
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87726001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGAACCGCGCCAGGGATCAGAG[A/C]ACCCAGAGTCCCCGCCCAGCTGCCG
Phenotype
MIM: 116880
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CTSL PubMed Links

Gene Details

Gene
CTSL
Gene Name
cathepsin L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257971.1 483 Intron NP_001244900.1
NM_001257972.1 483 Intron NP_001244901.1
NM_001257973.1 483 Intron NP_001244902.1
NM_001912.4 483 Intron NP_001903.1
NM_145918.2 483 Intron NP_666023.1
XM_005251716.3 483 UTR 5 XP_005251773.1
XM_011518263.1 483 UTR 5 XP_011516565.1
XM_017014293.1 483 UTR 5 XP_016869782.1

View Full Product Details