Product Details
- SNP ID
-
rs45550335
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132680929 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGTGCTTTTCAGACGTAACTATT[A/G]GGAGTGTATTTTGAAATACATATCA
- Phenotype
-
MIM: 603570
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VNN1
PubMed Links
Gene Details
- Gene
- VNN1
- Gene Name
- vanin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004666.2 |
3773 |
UTR 3 |
|
|
NP_004657.2 |
View Full Product Details