Product Details

SNP ID
rs41292894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:154157706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTGTTGCAAACCAAAGAAAAA[C/T]AGGTGGAAGGGCACTTTATGTCACA
Phenotype
MIM: 600018
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IPCEF1 PubMed Links

Gene Details

Gene
IPCEF1
Gene Name
interaction protein for cytohesin exchange factors 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130699.1 3836 UTR 3 NP_001124171.1
NM_001130700.1 3836 UTR 3 NP_001124172.1
NM_015553.2 3836 UTR 3 NP_056368.1
XM_005266919.4 3836 UTR 3 XP_005266976.1
XM_005266921.4 3836 UTR 3 XP_005266978.1
XM_005266923.3 3836 UTR 3 XP_005266980.1
XM_011535742.2 3836 UTR 3 XP_011534044.1
XM_011535744.2 3836 UTR 3 XP_011534046.1
XM_017010716.1 3836 UTR 3 XP_016866205.1
XM_017010717.1 3836 UTR 3 XP_016866206.1
XM_017010718.1 3836 UTR 3 XP_016866207.1
Gene
OPRM1
Gene Name
opioid receptor mu 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000914.4 3836 Intron NP_000905.3
NM_001008503.2 3836 Intron NP_001008503.2
NM_001008504.3 3836 Intron NP_001008504.2
NM_001008505.2 3836 Intron NP_001008505.2
NM_001145279.3 3836 Intron NP_001138751.1
NM_001145280.3 3836 Intron NP_001138752.1
NM_001145281.2 3836 Intron NP_001138753.1
NM_001145282.2 3836 Intron NP_001138754.1
NM_001145283.2 3836 Intron NP_001138755.1
NM_001145284.3 3836 Intron NP_001138756.1
NM_001145285.2 3836 Intron NP_001138757.1
NM_001145286.2 3836 Intron NP_001138758.1
NM_001145287.2 3836 Intron NP_001138759.1
NM_001285522.1 3836 Intron NP_001272451.1
NM_001285523.1 3836 Intron NP_001272452.1
NM_001285524.1 3836 Intron NP_001272453.1
NM_001285526.1 3836 Intron NP_001272455.1
NM_001285527.1 3836 Intron NP_001272456.1
NM_001285528.1 3836 Intron NP_001272457.1
XM_011535851.2 3836 Intron XP_011534153.1
XM_011535853.2 3836 Intron XP_011534155.1
XM_011535856.2 3836 Intron XP_011534158.1
XM_011535862.2 3836 Intron XP_011534164.1
XM_017010903.1 3836 Intron XP_016866392.1
XM_017010904.1 3836 Intron XP_016866393.1
XM_017010905.1 3836 Intron XP_016866394.1
XM_017010906.1 3836 Intron XP_016866395.1
XM_017010907.1 3836 Intron XP_016866396.1

View Full Product Details