Product Details

SNP ID
rs41299205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91114928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTGGGAGCATCAACACCAACA[C/T]GGCACAGGGGACTGCAGTGGTGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf159 PubMed Links

Gene Details

Gene
C14orf159
Gene Name
chromosome 14 open reading frame 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102366.2 294 Intron NP_001095836.1
NM_001102367.2 294 UTR 5 NP_001095837.1
NM_001102368.2 294 Intron NP_001095838.1
NM_001102369.2 294 Intron NP_001095839.1
NM_001286470.1 294 Intron NP_001273399.1
NM_001286471.1 294 UTR 5 NP_001273400.1
NM_001286472.1 294 Intron NP_001273401.1
NM_001286473.1 294 Intron NP_001273402.1
NM_024952.7 294 Intron NP_079228.4

View Full Product Details