Product Details

SNP ID
rs45600236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93240938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATTACACAACACCAAAATTTAG[A/T]CATATTTTGAAAATCGTTTAAAAGA
Phenotype
MIM: 610386
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BTBD7 PubMed Links

Gene Details

Gene
BTBD7
Gene Name
BTB domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002860.3 3845 UTR 3 NP_001002860.2
NM_001289133.1 3845 UTR 3 NP_001276062.1
NM_018167.4 3845 Intron NP_060637.1
XM_011536939.2 3845 Intron XP_011535241.1
XM_017021438.1 3845 Intron XP_016876927.1

View Full Product Details