Product Details

SNP ID
rs45546040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100826216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCCCACGAGAGGATCCCTCACC[C/T]GGTGAGTGGTTGGCCATCCTTGCCG
Phenotype
MIM: 605636
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEG3 PubMed Links

Gene Details

Gene
MEG3
Gene Name
maternally expressed 3 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details