Product Details

SNP ID
rs41497048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150053759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAAGTCAGTCCATTTCCATGAA[G/T]ATAAGGGGATTAGGAAAGAAGGTTC
Phenotype
MIM: 164770
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSF1R PubMed Links

Gene Details

Gene
CSF1R
Gene Name
colony stimulating factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288705.1 3521 UTR 3 NP_001275634.1
NM_005211.3 3521 UTR 3 NP_005202.2
Gene
HMGXB3
Gene Name
HMG-box containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014983.2 3521 Intron NP_055798.2
XM_006714765.2 3521 Intron XP_006714828.1

View Full Product Details