Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001270764.1 | 3340 | UTR 3 | NP_001257693.1 | ||
NM_001270765.1 | 3340 | Intron | NP_001257694.1 | ||
NM_014863.3 | 3340 | Intron | NP_055678.2 | ||
NM_015892.4 | 3340 | UTR 3 | NP_056976.2 | ||
XM_005269891.3 | 3340 | UTR 3 | XP_005269948.1 | ||
XM_005269892.4 | 3340 | Intron | XP_005269949.1 | ||
XM_005269893.3 | 3340 | Intron | XP_005269950.1 | ||
XM_005269894.4 | 3340 | Intron | XP_005269951.1 | ||
XM_006717891.3 | 3340 | UTR 3 | XP_006717954.1 | ||
XM_011539857.1 | 3340 | Intron | XP_011538159.1 | ||
XM_017016319.1 | 3340 | UTR 3 | XP_016871808.1 | ||
XM_017016320.1 | 3340 | Intron | XP_016871809.1 | ||
XM_017016321.1 | 3340 | Intron | XP_016871810.1 |