Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001276351.1 | 1361 | Silent Mutation | TAC,TAT | Y376Y | NP_001263280.1 |
NM_001276352.1 | 1361 | UTR 3 | NP_001263281.1 | ||
XM_011541465.2 | 1361 | Silent Mutation | TAC,TAT | Y447Y | XP_011539767.1 |
XM_011541466.2 | 1361 | Silent Mutation | TAC,TAT | Y447Y | XP_011539768.1 |
XM_011541467.1 | 1361 | Silent Mutation | TAC,TAT | Y434Y | XP_011539769.1 |
XM_011541469.1 | 1361 | Silent Mutation | TAC,TAT | Y309Y | XP_011539771.1 |
XM_011541472.1 | 1361 | Intron | XP_011539774.1 | ||
XM_011541473.2 | 1361 | Intron | XP_011539775.1 | ||
XM_011541475.1 | 1361 | Intron | XP_011539777.1 | ||
XM_017001276.1 | 1361 | Silent Mutation | TAC,TAT | Y447Y | XP_016856765.1 |
XM_017001277.1 | 1361 | Intron | XP_016856766.1 |