Product Details

SNP ID
rs41289228
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:75128469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAGCGGCTGGACCACTTGTGCT[C/G]GAGTGGTAAAGAACTATCATGAATC
Phenotype
MIM: 604425
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LHX8 PubMed Links

Gene Details

Gene
LHX8
Gene Name
LIM homeobox 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001933.1 36 UTR 5 NP_001001933.1
NM_001256114.1 36 Intron NP_001243043.1
XM_017001316.1 36 Intron XP_016856805.1
XM_017001317.1 36 Intron XP_016856806.1

View Full Product Details