Product Details

SNP ID
rs41289035
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:84814219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGATGCTCAGACCTTAGGGGAC[C/T]TATTCAGGTTTTCTTCTCTCCCAGC
Phenotype
MIM: 605106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPAR3 PubMed Links

Gene Details

Gene
LPAR3
Gene Name
lysophosphatidic acid receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012152.2 Intron NP_036284.1

View Full Product Details