Product Details

SNP ID
rs41465250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62422264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCTTTCAGCGTGTCATCGAAAC[C/T]CTCCTCATGGACACACCCTCCAGTT
Phenotype
MIM: 192020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCGB1A1 PubMed Links

Gene Details

Gene
SCGB1A1
Gene Name
secretoglobin family 1A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003357.4 160 Silent Mutation ACC,ACT T33T NP_003348.1

View Full Product Details